
5,7-Bis(trifluoromethyl)-4-hydroxyquinoline-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:240408-95-1
:
Formula:C₁₂H₅F₆NO₃
MDL Number:MFCD00269347
:325.163 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together