


Trifluoroborane acetonitrile complex solution
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:420-16-6
:
Formula:BF₃·CH₃CN
MDL Number:MFCD28125563
:108.858 g/mol
Density:0.87 g/cm³ (25 °C)
Flash Pt:10 °C
UN:2924
ADR:3+8,II
:GHS02
:GHS05
:GHS06
Storage Temperature:Refrigerator
Frequently Bought Together