2-(4-Bromophenyl)-2-[4-(4-fluorobenzoyl)phenyl]acetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:339115-52-5
:
Formula:C₂₁H₁₃BrFNO
MDL Number:MFCD00792170
:394.236 g/mol
Melting Pt:136…138 °C
Storage Temperature:Ambient
Frequently Bought Together