
(R)-3,3-Difluoropropane-1,2-diol 98% (85%ee)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1026908-13-3
:
Formula:C₃H₆F₂O₂
MDL Number:MFCD29988691
:112.075 g/mol
Boiling Pt:113 °C (37.5 mm Hg)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together