Order Entry
Austria
ContactUsLinkComponent
 
[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile
Catalog # APOSPC410476-1G
Supplier:  Apollo Scientific
CAS Number:  
[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile
Catalog # APOSPC410476-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :209.556 g/mol
Storage Temperature:Ambient
CAS Number:1006471-49-3
Formula:C₆H₃ClF₃N₃
MDL Number:MFCD04969710