Order Entry
Austria
ContactUsLinkComponent
1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410325-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410325-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :208.138 g/mol
Storage Temperature:Ambient
CAS Number:1004643-70-2
Formula:C₇H₇F₃N₂O₂
MDL Number:MFCD04970207