Order Entry
Austria
ContactUsLinkComponent
1-(Difluoromethyl)-3,5-dimethyl-1H-pyrazol-4-amine hydrochloride
1-(Difluoromethyl)-3,5-dimethyl-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410114-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(Difluoromethyl)-3,5-dimethyl-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410114-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.614 g/mol
Storage Temperature:Ambient
Formula:C₆H₉F₂N₃·ClH
MDL Number:MFCD18089461