Order Entry
Austria
ContactUsLinkComponent
3-{[(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]sulphanyl}-5-(trifluoromethyl)-4H-1,2,4-triazol-4-amine
3-{[(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]sulphanyl}-5-(trifluoromethyl)-4H-1,2,4-triazol-4-amine
Catalog # APOSPC410021-1G
Supplier:  Apollo Scientific
CAS Number:  
3-{[(3,5-Dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]sulphanyl}-5-(trifluoromethyl)-4H-1,2,4-triazol-4-amine
Catalog # APOSPC410021-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :337.282 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₀F₃N₇O₂S
MDL Number:MFCD05861715