5,7-Difluoro-4-hydroxyquinoline
Catalog # APOSPC404580-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:181.139 g/mol
Storage Temperature:Ambient
CAS Number:874804-43-0
Formula:C₉H₅F₂NO
MDL Number:MFCD06409136