Order Entry
Austria
ContactUsLinkComponent
7,8-Difluoro-4-hydroxyquinoline
7,8-Difluoro-4-hydroxyquinoline
Catalog # APOSPC404577-1G
Supplier:  Apollo Scientific
CAS Number:  
7,8-Difluoro-4-hydroxyquinoline
Catalog # APOSPC404577-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :181.139 g/mol
Storage Temperature:Ambient
CAS Number:1142193-11-0
Formula:C₉H₅F₂NO
MDL Number:MFCD11870786