Order Entry
Austria
ContactUsLinkComponent
4-Bromo-5-fluorobenzene-1,2-diol
4-Bromo-5-fluorobenzene-1,2-diol
Catalog # APOSPC402028-250MG
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-5-fluorobenzene-1,2-diol
Catalog # APOSPC402028-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :206.997 g/mol
Storage Temperature:Ambient
CAS Number:656804-73-8
Formula:C₆H₄BrFO₂
MDL Number:MFCD13177846