
Methyl 3,6-difluoro-2-(trifluoromethoxy)benzoate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2366994-17-2
:
Formula:C₉H₅F₅O₃
MDL Number:MFCD32066742
:256.126 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together