Ethyl 6-chloro-8-fluoro-4-hydroxyquinoline-3-carboxylate, tech
Catalog # APOSPC28115-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:269.656 g/mol
Storage Temperature:Ambient
CAS Number:338795-16-7
Formula:C₁₂H₉ClFNO₃
MDL Number:MFCD00173397