Order Entry
Austria
ContactUsLinkComponent
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetonitrile 95%
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetonitrile 95%
Catalog # APOSPC28054-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[2-(Trifluoromethyl)-1H-1,3-benzodiazol-1-yl]acetonitrile 95%
Catalog # APOSPC28054-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.17 g/mol
Storage Temperature:Ambient
CAS Number:478042-43-2
Formula:C₁₀H₆F₃N₃
MDL Number:MFCD02187083