2-{3-[(4-Fluorophenyl)methyl]-4-oxo-3,4-dihydrophthalazin-1-yl}acetonitrile, tech
Catalog # APOSPC28053-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:293.295 g/mol
Storage Temperature:Ambient
CAS Number:478041-35-9
Formula:C₁₇H₁₂FN₃O
MDL Number:MFCD02186453