Order Entry
Austria
ContactUsLinkComponent
Ethyl 5-(3,4-difluorophenyl)-1-methyl-1H-pyrazole-3-carboxylate

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :266.243 g/mol
Storage Temperature:Ambient
CAS Number:1170363-38-8
Formula:C₁₃H₁₂F₂N₂O₂
MDL Number:MFCD14522979