8-Bromo-6-fluoro-4-hydroxyquinoline-3-carboxylic acid
Catalog # APOSPC110035-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:286.054 g/mol
Storage Temperature:Ambient
CAS Number:1019016-15-9
Formula:C₁₀H₅BrFNO₃
MDL Number:MFCD09861322