3,5-Dimethyl-4-(hydroxymethyl)-1-phenyl-1H-pyrazole
Catalog # APOSOR9983-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.252 g/mol
Melting Pt:106…110 °C
Storage Temperature:Ambient
CAS Number:58789-53-0
Formula:C₁₂H₁₄N₂O
MDL Number:MFCD06202725