Order Entry
Austria
ContactUsLinkComponent
4-tert-Butyl 2-ethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate 98%
4-tert-Butyl 2-ethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate 98%
Catalog # APOSOR9937-1G
Supplier:  Apollo Scientific
CAS Number:  
4-tert-Butyl 2-ethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate 98%
Catalog # APOSOR9937-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3-(tert-Butoxycarbonyl)-2,4-dimethyl-5-(ethoxycarbonyl)-1H-pyrrole
 :
 :267.321 g/mol
Melting Pt:125…127 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:361380-77-0
Formula:C₁₄H₂₁NO₄
MDL Number:MFCD03093037