Ethyl 1-methyl-3-phenyl-1H-pyrazole-5-carboxylate 95+%
Catalog # APOSOR9786-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1-[5-(Ethoxycarbonyl)-1-methyl-1H-pyrazol-3-yl]benzene, 5-(Ethoxycarbonyl)-1-methyl-3-phenyl-1H-pyrazole
:
:230.262 g/mol
Melting Pt:27 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:10250-63-2
Formula:C₁₃H₁₄N₂O₂
MDL Number:MFCD03933288