
3-(Aminomethyl)benzaldehyde, N-BOC protected 90%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:170853-04-0
:
Formula:C₁₃H₁₇NO₃
MDL Number:MFCD06659091
:235.279 g/mol
Melting Pt:59 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together