
1,3-Dimethyl-1H-pyrazole-5-thiocarboxamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:844891-03-8
:
Formula:C₆H₉N₃S
MDL Number:MFCD06659061
:155.221 g/mol
Melting Pt:170 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together