Order Entry
Austria
ContactUsLinkComponent
1-(3-Hydroxyphenyl)-3-methylbutan-1-one 95%
1-(3-Hydroxyphenyl)-3-methylbutan-1-one 95%
Catalog # APOSOR957667-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Hydroxyphenyl)-3-methylbutan-1-one 95%
Catalog # APOSOR957667-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :178.228 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₁₄O₂
MDL Number:MFCD19313738