(6,7,8,9-Tetrahydro-5h-[1,2,4]triazolo[4,3-a]azepin-3-yl)-acetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:116598-69-7
:
Formula:C₉H₁₂N₄
MDL Number:MFCD00980781
:176.218 g/mol
Storage Temperature:Ambient
Frequently Bought Together