Order Entry
Austria
ContactUsLinkComponent
3,6-Dimethyl-2-nitrophenol 95%
3,6-Dimethyl-2-nitrophenol 95%
Catalog # APOSOR951519-1G
Supplier:  Apollo Scientific
CAS Number:  
3,6-Dimethyl-2-nitrophenol 95%
Catalog # APOSOR951519-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.162 g/mol
Storage Temperature:Ambient
CAS Number:71608-10-1
Formula:C₈H₉NO₃
MDL Number:MFCD12198862