Order Entry
Austria
ContactUsLinkComponent
Octa-3,5-diyne-1,8-diol 95%
Octa-3,5-diyne-1,8-diol 95%
Catalog # APOSOR946829-1G
Supplier:  Apollo Scientific
CAS Number:  
Octa-3,5-diyne-1,8-diol 95%
Catalog # APOSOR946829-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :138.164 g/mol
Storage Temperature:Ambient
CAS Number:15808-23-8
Formula:C₈H₁₀O₂
MDL Number:MFCD00190210