Order Entry
Austria
ContactUsLinkComponent
(Tetrahydro-pyran-2-yloxy)-acetonitrile 95%
(Tetrahydro-pyran-2-yloxy)-acetonitrile 95%
Catalog # APOSOR944255-1G
Supplier:  Apollo Scientific
CAS Number:  
(Tetrahydro-pyran-2-yloxy)-acetonitrile 95%
Catalog # APOSOR944255-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :141.168 g/mol
Storage Temperature:Ambient
CAS Number:17521-49-2
Formula:C₇H₁₁NO₂
MDL Number:MFCD21362389