Order Entry
Austria
ContactUsLinkComponent
Ethyl 1-methyl-1H-pyrazole-5-carboxylate 95%
Ethyl 1-methyl-1H-pyrazole-5-carboxylate 95%
Catalog # APOSOR939418-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 1-methyl-1H-pyrazole-5-carboxylate 95%
Catalog # APOSOR939418-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

 :
 :154.166 g/mol
Storage Temperature:Ambient
CAS Number:197079-26-8
Formula:C₇H₁₀N₂O₂
MDL Number:MFCD08700760