2-(Azetidin-3-yl)acetonitrile hydrochloride
Artikelnr. APOSOR933553-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:132.591 g/mol
Storage Temperature:Ambient
CAS Number:1423057-36-6
Formula:C₅H₈N₂·ClH
MDL Number:MFCD28501222