2-(1-(Ethylsulphonyl)azetidin-3-ylidene)acetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1187595-85-2
:
Formula:C₇H₁₀N₂O₂S
MDL Number:MFCD22987586
:186.232 g/mol
Storage Temperature:Ambient
Frequently Bought Together