Order Entry
Austria
ContactUsLinkComponent
 
(R)-2-Methylbutane-2-sulphinamide 95%
Catalog # APOSOR916267-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-2-Methylbutane-2-sulphinamide 95%
Catalog # APOSOR916267-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :135.228 g/mol
Storage Temperature:Ambient
CAS Number:446021-71-2
Formula:C₅H₁₃NOS
MDL Number:MFCD11707200