(6-(1H-Pyrazol-1-yl)pyridin-3-yl)methanol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:748796-38-5
:
Formula:C₉H₉N₃O
MDL Number:MFCD06658507
:175.187 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together