Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: Ethyl 3-cyclopropyl-1H-pyrazole-5-carboxylate, Ethyl 5-cyclopropyl-1H-pyrazole-3-carboxylat
:
:180.204 g/mol
Melting Pt:90…92 °C
Storage Temperature:Refrigerator
CAS Number:133261-06-0
Formula:C₉H₁₂N₂O₂
MDL Number:MFCD16658044