Order Entry
Austria
ContactUsLinkComponent
3-Propoxypropane-1,2-diol
Irritant
3-Propoxypropane-1,2-diol
Catalog # APOSOR60027-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Propoxypropane-1,2-diol
Catalog # APOSOR60027-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:129…130 °C (25 mm Hg)
Density:1.0244 g/cm³ (25 °C)
 :134.174 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:61940-71-4
Formula:C₆H₁₄O₃
MDL Number:MFCD00128145