
5-Bromo-4-chloro-2-nitrobenzamide 98%
Catalog # APOSOR52444-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:279.475 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1820608-87-4
Formula:C₇H₄BrClN₂O₃
MDL Number:MFCD28127174