1-Azido-1-deoxy-β-D-glucopyranoside tetraacetate 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl azide, β-D-Glucopyranosyl azide tetraacetate
CAS Number:13992-25-1
:
Formula:C₁₄H₁₉N₃O₉
MDL Number:MFCD00216966
:373.315 g/mol
Melting Pt:122…124 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together