(1-Cyclopropyl-1H-pyrazol-4-yl)methanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1780993-51-2
:
Formula:C₇H₁₀N₂O
MDL Number:MFCD28550088
:138.167 g/mol
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together