
2-(Azetidin-3-ylidene)acetonitrile (hydrochloride) 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1314910-43-4
:
Formula:C₅H₆N₂·ClH
MDL Number:MFCD19227479
:130.575 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together