Order Entry
Austria
ContactUsLinkComponent
(E)-2-(1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetonitrile ≥98%
(E)-2-(1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetonitrile ≥98%
Catalog # APOSOR370075-1G
Supplier:  Apollo Scientific
CAS Number:  
(E)-2-(1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetonitrile ≥98%
Catalog # APOSOR370075-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.233 g/mol
Storage Temperature:Ambient
CAS Number:196597-79-2
Formula:C₁₃H₁₁NO
MDL Number:MFCD09955086