1-(3-Chloro-phenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:294877-36-4
:
Formula:C₁₂H₁₁ClN₂O
MDL Number:MFCD09939992
:234.682 g/mol
Storage Temperature:Ambient
Frequently Bought Together