2-(3,5-Dimethyl-4-propyl-1H-pyrazol-1-yl)pyridin-3-amine, tech
Catalog # APOSOR32586-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:230.309 g/mol
Storage Temperature:Ambient
CAS Number:955966-83-3
Formula:C₁₃H₁₈N₄
MDL Number:MFCD03787559