5,6-Dibromocyclohex-2-ene-1,4-diol, tech
Catalog # APOSOR32443-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.935 g/mol
Storage Temperature:Ambient
CAS Number:317821-92-4
Formula:C₆H₈Br₂O₂
MDL Number:MFCD01568911