2-Ethyl 5-(4-nitrophenyl)methyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyrazine-2,5-dicarboxylate
Artikelnr. APOSOR310410-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:374.348 g/mol
Storage Temperature:Ambient
CAS Number:623565-14-0
Formula:C₁₇H₁₈N₄O₆
MDL Number:MFCD27924230