Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Bromo-2,8-dimethyl-4-hydroxyquinoline
7-Bromo-2,8-dimethyl-4-hydroxyquinoline
7-Bromo-2,8-dimethyl-4-hydroxyquinoline
APOSOR309315-1G
:
Apollo Scientific
:
7-Bromo-2,8-dimethyl-4-hydroxyquinoline
APOSOR309315-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
252.107 g/mol
:
Ambient
:
1189106-80-6
:
C₁₁H₁₀BrNO
:
MFCD12675041