Order Entry
Austria
ContactUsLinkComponent
7-Chloro-2-hydroxyquinoline-3-carbaldehyde
7-Chloro-2-hydroxyquinoline-3-carbaldehyde
Catalog # APOSOR309255-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Chloro-2-hydroxyquinoline-3-carbaldehyde
Catalog # APOSOR309255-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :207.613 g/mol
Storage Temperature:Ambient
CAS Number:73568-43-1
Formula:C₁₀H₆ClNO₂
MDL Number:MFCD09881442