Order Entry
Austria
ContactUsLinkComponent
6-Chloro-2-hydroxyquinoline-3-carbaldehyde
6-Chloro-2-hydroxyquinoline-3-carbaldehyde
Catalog # APOSOR309254-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-2-hydroxyquinoline-3-carbaldehyde
Catalog # APOSOR309254-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :207.613 g/mol
Storage Temperature:Ambient
CAS Number:73568-44-2
Formula:C₁₀H₆ClNO₂
MDL Number:MFCD09881441