Order Entry
Austria
ContactUsLinkComponent
6,7-Dimethyl-4-hydroxyquinoline
6,7-Dimethyl-4-hydroxyquinoline
Catalog # APOSOR307996-1G
Supplier:  Apollo Scientific
CAS Number:  
6,7-Dimethyl-4-hydroxyquinoline
Catalog # APOSOR307996-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :173.211 g/mol
Storage Temperature:Ambient
CAS Number:185437-33-6
Formula:C₁₁H₁₁NO
MDL Number:MFCD00272457