Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxyquinoline-5-carboxylic acid
2-Hydroxyquinoline-5-carboxylic acid
2-Hydroxyquinoline-5-carboxylic acid
APOSOR302189-1G
:
Apollo Scientific
:
2-Hydroxyquinoline-5-carboxylic acid
APOSOR302189-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
189.167 g/mol
:
Ambient
:
83734-43-4
:
C₁₀H₇NO₃
:
MFCD11557267