6-Methoxy-4-hydroxyquinoline
Catalog # APOSOR301003-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:175.184 g/mol
Storage Temperature:Ambient
CAS Number:23432-39-5
Formula:C₁₀H₉NO₂
MDL Number:MFCD00014662