Order Entry
Austria
ContactUsLinkComponent
 
Irritant
Bis(5-chloro-1,3-dimethyl-1H-pyrazol-4-yl)methanol
Catalog # APOSOR24176-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Bis(5-chloro-1,3-dimethyl-1H-pyrazol-4-yl)methanol
Catalog # APOSOR24176-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :289.161 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:648409-46-5
Formula:C₁₁H₁₄Cl₂N₄O
MDL Number:MFCD00121975